Learn More
Al-23 Tube, Both Ends Open, O.D.: 5mm, I.D.: 3mm, Thermo Scientific™
Product Code. 11333927
Change view
Click to view available options
Quantity:
100 mm
200 mm
300 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
300mm
600mm
800mm
Product Code. Quantity unitSize
11333927 200 mm 200mm
11393917 100 mm 100mm
11313927 300 mm 300mm
11303927 600 mm 600mm
11353927 800 mm 800mm
11323927 1000 mm 1000mm
6 options
Product Code. 11333927
Brand: Thermo Scientific Alfa Aesar 031555.DR
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
Quantity 200 mm