Learn More
Al-23 Solid Rod, Round;Diameter (mm), 5
Product Code. 11363078
Change view
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
400 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
250mm
300mm
400mm
600mm
800mm
Product Code. Quantity unitSize
11363078 200 mm 200mm
11343078 100 mm 100mm
11353078 250 mm 250mm
11333078 300 mm 300mm
15421297 400 mm 400mm
11303088 600 mm 600mm
11383078 800 mm 800mm
11393078 1000 mm 1000mm
8 options
Product Code. 11363078
Brand: Thermo Scientific Alfa Aesar 032931.DR
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Solid Rod
Quantity 200 mm
Form Rod
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.