Learn More
Al-23 Solid Rod, Round, Dia.: 3mm, Thermo Scientific™
Product Code. 11373058
Change view
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
250mm
300mm
600mm
800mm
Product Code. Quantity unitSize
11373058 100 mm 100mm
11333068 200 mm 200mm
11383058 250 mm 250mm
11343068 300 mm 300mm
11393058 600 mm 600mm
11313068 800 mm 800mm
11323068 1000 mm 1000mm
7 options
Product Code. 11373058
Brand: Thermo Scientific Alfa Aesar 032927.DM
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Solid Rod
Quantity 100 mm
Form Rod