Learn More
Al-23 Solid Rod, Round, Dia.: 1.5mm, Thermo Scientific™
Product Code. p-4905258
Change view
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
Unit Size:
100mm
200mm
250mm
300mm
Product Code. Quantity unitSize
11303058 100 mm 100mm
11373048 200 mm 200mm
11393048 250 mm 250mm
11383048 300 mm 300mm
4 options
Product Code. 11303058
Brand: Thermo Scientific Alfa Aesar 032925.DM
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Solid Rod
Quantity 100 mm
Form Rod