Learn More
Al-23 Protective Tube, One End Closed, O.D.: 12mm, I.D.: 8mm, Wall.: 2mm, Thermo Scientific™
Product Code. 11342688
Change view
Click to view available options
Quantity:
200 mm
270 mm
375 mm
530 mm
740 mm
1030 mm
1460 mm
2030 mm
Unit Size:
1030mm
1460mm
200mm
2030mm
270mm
375mm
530mm
740mm
Product Code. Quantity unitSize
11342688 200 mm 200mm
11322688 270 mm 270mm
11372678 375 mm 375mm
11302688 530 mm 530mm
11392678 740 mm 740mm
11332688 1030 mm 1030mm
15471137 1460 mm 1460mm
11382678 2030 mm 2030mm
8 options
Product Code. 11342688
Brand: Thermo Scientific Alfa Aesar 030048.DR
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Protective Tube
Name Note One end closed
Quantity 200 mm