Learn More
Al-23 Multibore Round Tube, O.D.: 2.8mm, Four 0.7mm Bores, Thermo Scientific™
Product Code. 11318527
Change view
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
Unit Size:
100mm
200mm
250mm
300mm
Product Code. Quantity unitSize
11318527 250 mm 250mm
11308527 100 mm 100mm
11388517 200 mm 200mm
11378517 300 mm 300mm
4 options
Product Code. 11318527
Brand: Thermo Scientific Alfa Aesar 032603.DT
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Multibore Round Tube
Name Note Four bores
Quantity 250 mm