Learn More
Al-23 Multibore Round Tube, O.D.: 1.2mm, Two 0.3mm Bores, Thermo Scientific Chemicals
Product Code. 11398467
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
400 mm
Unit Size:
100mm
200mm
250mm
300mm
400mm
Product Code. 11398467
Brand: Thermo Scientific Alfa Aesar 032565.DW
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Multibore Round Tube
Name Note Two bores
Quantity 300 mm