Learn More
Al-23 Multibore Round Tube, O.D.: 1.2mm, Two 0.3mm Bores, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
400 mm
Unit Size:
100mm
200mm
250mm
300mm
400mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Al-23 Multibore Round Tube |
| Name Note | Two bores |
| Quantity | 100 mm |