Learn More
Al-23 Multibore Insulating Tube, O. D.: 8mm, Two 1.6mm Bores, Thermo Scientific™
Product Code. 11358447
Change view
Click to view available options
Quantity:
205 mm
275 mm
380 mm
560 mm
770 mm
1060 mm
1460 mm
2060 mm
Unit Size:
1060mm
1460mm
205mm
2060mm
275mm
380mm
560mm
770mm
Product Code. Quantity unitSize
11358447 1060 mm 1060mm
11328447 205 mm 205mm
11388447 275 mm 275mm
11378447 380 mm 380mm
11348447 560 mm 560mm
11338447 770 mm 770mm
11398447 1460 mm 1460mm
11368447 2060 mm 2060mm
8 options
Product Code. 11358447
Brand: Thermo Scientific Alfa Aesar 032558.ER
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Multibore Insulating Tube
Name Note Two bores
Quantity 1060 mm