Learn More
Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1.6;ID (mm), 0.8
Product Code. 15441187
Change view
Click to view available options
Quantity:
100 mm
200 mm
300 mm
400 mm
Unit Size:
100mm
200mm
300mm
400mm
Product Code. Quantity unitSize
15441187 400 mm 400mm
11397498 100 mm 100mm
11307508 200 mm 200mm
11377498 300 mm 300mm
4 options
Product Code. 15441187
Brand: Thermo Scientific Alfa Aesar 031485.Q@
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Insulating Tube
Name Note Both ends open (Thin Wall)
Quantity 400 mm
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.