Learn More
Al-23 Insulating Beads, O.D.: 4mm, Four 0.8mm Bores, Length: 4mm, Thermo Scientific™
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Al-23 Insulating Beads |
| Name Note | 4 channels |
| Quantity | 100 Pk. |
| Form | Insulating Beads |