missing translation for 'onlineSavingsMsg'
Learn More

AKTIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14860-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893724

  • 4593.76 NOK / 150µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

Component of the FTS/Hook/FHIP complex (FHF complex). The FHF complex may function to promote vesicle trafficking and/or fusion via the homotypic vesicular protein sorting complex (the HOPS complex). Regulates apoptosis by enhancing phosphorylation and activation of AKT1. Increases release of TNFSF6 via the AKT1/GSK3B/NFATC1 signaling cascade.
Specifications

Specifications

AKTIP
Polyclonal
Unconjugated
AKTIP
AKT interacting protein, AKTIP, FT1, FTS, Fused toes protein homolog
Rabbit
Antigen Affinity Chromatography
RUO
14339, 291906, 64400
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5FVH4, Q64362, Q9H8T0
Aktip
AKTIP Fusion Protein Ag6662
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers