missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKTIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14860-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Component of the FTS/Hook/FHIP complex (FHF complex). The FHF complex may function to promote vesicle trafficking and/or fusion via the homotypic vesicular protein sorting complex (the HOPS complex). Regulates apoptosis by enhancing phosphorylation and activation of AKT1. Increases release of TNFSF6 via the AKT1/GSK3B/NFATC1 signaling cascade.Specifications
AKTIP | |
Polyclonal | |
Unconjugated | |
AKTIP | |
AKT interacting protein, AKTIP, FT1, FTS, Fused toes protein homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14339, 291906, 64400 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5FVH4, Q64362, Q9H8T0 | |
Aktip | |
AKTIP Fusion Protein Ag6662 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |