missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ AKR7A3 Recombinant Protein

Brand:  Abnova™ H00022977-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16146812

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH25709
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
61.93
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKR7A3
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
22977
AKR7A3 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
AFAR2
AKR7A3
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers