missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR7A2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10730-1-AP-150UL
This item is not returnable.
View return policy
Description
AKR7A2 is aldo-keto reductases, which are involved in the detoxification of aldehydes and ketones.Specifications
| AKR7A2 | |
| Polyclonal | |
| Unconjugated | |
| AKR7A2 | |
| AFAR, AFAR1, AFB1 aldehyde reductase 1, AFB1 AR 1, AFB1 AR1, AKR7, AKR7A2, Aldoketoreductase 7, SSA reductase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110198, 8574 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43488, Q8CG76 | |
| AKR7A2 | |
| AKR7A2 Fusion Protein Ag1049 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction