missing translation for 'onlineSavingsMsg'
Learn More

AKR7A2 Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™

Product Code. 30499747 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499747 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499747 Supplier Novus Biologicals Supplier No. NBP335236JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AKR7A2 Polyclonal antibody specifically detects AKR7A2 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR7A2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias AFAR1, AFARAFB1-AR1, AFB1 aldehyde reductase 1, AFB1-AR 1, aflatoxin B1 aldehyde reductase member 2, aflatoxin beta1 aldehyde reductase, AKR7, aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase), Aldoketoreductase 7, EC 1.1.1.n11, SSA reductase, Succinic semialdehyde reductase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-359 of human AKR7A2 (NP_003680.2).,, Sequence:, VKIATKANPWDGKSLKPDSVRSQLETSLKRLQCPQVDLFYLHAPDHGTPVEETLHACQRLHQEGKFVELGLSNYASWEVAEICTLCKSNGWILPTVYQGMYNATTRQVETELFPCLRHFGLRFYAYNPLAGGLLTGKYKYEDKDGKQPVGRFFGNSWAETYRNRFWKEHHFEAIALVEKALQAAYGASAPSVTSAALRWMYHHSQLQGAHGDAVILGMSSLEQLEQNLAATEEGPLEPAVVDAFNQAWHLVAHECPNYFR
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8574
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.