missing translation for 'onlineSavingsMsg'
Learn More

AKR1C4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18852-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888704

  • 3468.15 NOK / 150µL
Estimated Shipment: 19-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the bioreduction of chlordecone, a toxic organochlorine pesticide, to chlordecone alcohol in liver. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14.
Specifications

Specifications

AKR1C4
Polyclonal
Unconjugated
AKR1C4
3 alpha HSD, 3 alpha HSD1, AKR1C4, C11, CDR, CHDR, Chlordecone reductase, DD 4, DD4, Dihydrodiol dehydrogenase 4, HAKRA
Rabbit
Antigen Affinity Chromatography
RUO
1109
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P17516
AKR1C4
AKR1C4 Fusion Protein Ag13444
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers