missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1C4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18852-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the bioreduction of chlordecone, a toxic organochlorine pesticide, to chlordecone alcohol in liver. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14.Specifications
AKR1C4 | |
Polyclonal | |
Unconjugated | |
AKR1C4 | |
3 alpha HSD, 3 alpha HSD1, AKR1C4, C11, CDR, CHDR, Chlordecone reductase, DD 4, DD4, Dihydrodiol dehydrogenase 4, HAKRA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1109 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17516 | |
AKR1C4 | |
AKR1C4 Fusion Protein Ag13444 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |