missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AKR1B1 (Human) Recombinant Protein
Human AKR1B1 full-length ORF ( AAH00260.1, 1 a.a. - 316 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000231-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MASRLLLNNGAKMPILGLGTWKSPPGQVTEAVKVAIDVGYRHIDCAHVYQNENEVGVAIQEKLREQVVKREELFIVSKLWCTYHEKGLVKGACQKTLSDLKLDYLDLYLIHWPTGFKPGKEFFPLDESGNVVPSDTNILDTWAAMEELVDEGLVKAIGISNFNHLQVEMILNKPGLKYKPAVNQIECHPYLTQEKLIQYCQSKGIVVTAYSPLGSPDRPWAKPEDPSLLEDPRIKAIAAKHNKTTAQVLIRFPMQRNLVVIPKSVTPERIAENFKVFDFELSSQDMTTLLSYNRNWRVCALLSCTSHKDYPFHEEF
Specifications
AAH00260.1 | |
aldo-keto reductase family 1, member B1 (aldose reductase) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AKR1B1 | |
GST |
231 | |
Wheat germ expression system | |
10 μg | |
ADR, ALDR1, ALR2, AR, MGC1804 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |