Learn More
Honeywell™ Airvisor Carbon Cartridge
Product Code. 12662663
Change view
Click to view available options
Applications:
Removing Odor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Applications Unit Size
12662663 Removing Odor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12662663 Supplier Honeywell™ Supplier No. 1001672
  • ONLINE EXCLUSIVE PRICE
    1176.00 NOK
    1045.42 NOK / Pack of 1
    Save 130.58 NOK
    11% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Carbon filter suitable for all Airvisor air respirators.
  • Suitable for all Airvisor models
  • Removes unpleasant odor
  • Should be replaced by 1000 hours or when unpleasant smell is experienced
  • Comes with two replacement O-rings
  • Part of an air respirator system certified to EN14594
TRUSTED_SUSTAINABILITY
Specifications
Applications Removing Odor
For Use With (Equipment) Airvisor Air Respirators