Description
- Heads and cameras for use with 3M™ Airstream helmet
- For use with 3M™ Airstream helmets
- Genuine 3M™ accessories
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | Airstream™ Powered Air Respirator System |
| Product Type | Pivot and Cams Set |