Learn More
Syngene™ AirClean™ Systems Dead Air Box with UVTect™ Controller
GREENER_CHOICE
Product Code. 17725904 Shop All Syngene Products
Change view
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) unitSize
17725904 32 in. Each
17795904 48 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17725904 Supplier Syngene™ Supplier No. SYNAC632TDBC220E
  • ONLINE EXCLUSIVE PRICE
    84780.00 NOK
    74862.19 NOK / Pack of 1
    Save 9917.81 NOK
    11% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
The AirClean™ Systems Dead Air Box provides a circulation-free environment for the amplification and manipulation of DNA/RNA. Each dead air box is standard with the UVTect™ Microprocessor Controller and a 254 nm ultraviolet light source for irradiation between amplifications.
AirClean™ Systems Dead Air Boxes provide a circulation-free environment for the amplification and manipulation of DNA/RNA. Each dead air box is standard with the UVTect™ Microprocessor Controller and a 254 nm ultraviolet light source for irradiation between amplifications. A safety interlock switch disables the UV light when the sash is raised, preventing operator exposure. Seamless design and UV-reflective construction materials further ensure no harmful UV energy escapes the enclosure.
Standard Features
  • Polycarbonate and polypropylene design reflects UV energy
  • Shipped fully assembled
  • Overlapping polycarbonate sash to prevent UV light exposure
  • Sash safety switch automatically turns off UV light when sash is raised
  • Electronics are located above the workspace, eliminating the chance of a chemical spill entering these sensitive areas
  • Unique Safety Features
  • No ductwork required
  • No installation required - plugs directly into a standard 110V or 220V electrical outlet
  • UVTect™ Microprocessor Controller
  • UV and fluorescent lights
  • Seamless plastic design -no joints or gaps in construction
  • UV shelf with built-in pipettor holder
  • Lab event timer
  • Digital UV light timer 0.01 seconds to 59 minutes
  • Access port for electrical cords
Specifications
Product Type Dead Air Box
For Use With (Equipment) Standard/Tall Polycarbonate UV Units
Includes UVTect™ Microprocessor Controller, 254 nm Ultraviolet Light Source
Model Tall Version
Width (English) 32 in.
Width (Metric) 81 cm