Learn More
Syngene™ Airclean™ Process Protection Microscope Hood
GREENER_CHOICE
Product Code. 16532972 Shop All Syngene Products
Change view
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) unitSize
16532972 32 in. Each
16552972 48 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16532972 Supplier Syngene™ Supplier No. SYN32PPUVCE
  • ONLINE EXCLUSIVE PRICE
    111340.00 NOK
    98317.68 NOK / Pack of 1
    Save 13022.32 NOK
    11% Off
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Process Protection with UVTect™ Microprocessor and Microscope Sash
  • AirClean™ Systems AC600 Series Vertical Laminar Flow Workstation
  • Laminar air flow is described as the smooth, fluid, parallel flow of air in a consistent direction
  • Ideal for applications that are not hazardous to the operator but require a clean workspace
  • A compact, light-weight design allows for multiple clean zones within a laboratory
  • Two-stage filtration provides clean, Class 100 laminar flow air for process protection
  • Room air first passes through an electrostatically charged pre-filter followed by a HEPA filter resulting in ISO 5 clean air
  • Independently certified to meet or exceed applicable clean bench standards
Specifications
Product Type Laminar Flow
Depth (Metric) 61 cm
Height (Metric) 76.2 cm
Width (English) 32 in.
Width (Metric) 81.28 cm