Learn More
Syngene™ Airclean™ Operator Protection Microscope Sash
GREENER_CHOICE
Product Code. 16522972 Shop All Syngene Products
Change view
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) unitSize
16522972 32 in. Each
16542972 48 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16522972 Supplier Syngene™ Supplier No. SYN32OPASE
  • ONLINE EXCLUSIVE PRICE
    113240.00 NOK
    99993.06 NOK / Pack of 1
    Save 13246.94 NOK
    11% Off
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
Operator Protection Enclosure fitted with AirSafe™ Automatic Safety Controller
  • Ductless microscope enclosures provide an ISO 5 work environment for sample preparation
  • Downflow microscope enclosure provides a clean environment with UV light for irradiation between processes
  • UVTect™ microprocessor controller provides one-touch operation of all enclosure features
  • Each enclosure is designed to work with a wide range of microscopes
  • Access port provides easy access to the eye piece and viewing platform
  • Configuration does not provide operator protection
Specifications
Product Type Laminar Flow
Depth (Metric) Interior 61 cm
Height (Metric) 101.6 cm
Width (English) 32 in.
Width (Metric) 81.28 cm