Learn More
Syngene™ Airclean 600 PCR Tall Workstation
Product Code. 16564341 Shop All Syngene Products
Change view
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Unit Size
16564341 48 in. Each
16524341 32 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16564341 Supplier Syngene™ Supplier No. SYN48PWTLFE
  • ONLINE EXCLUSIVE PRICE
    134110.00 NOK
    118422.42 NOK / Pack of 1
    Save 15687.58 NOK
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-01-2027
Airclean 600 PCR Tall Workstation with UVTect microprocessor controller and vertical laminar air flow.
  • Tall version
  • With UVTect microprocessor controller
  • Provides Class 100 vertical laminar air flow and timed uv light for sterilization
  • Includes HEPA filter, initial prefilter, fluorescent light, UV shelf with pipette holder, safety switch, and slip hatch
Specifications
Product Type Tall Workstation
Certifications/Compliance UL, CSA, and CE approved
Includes UV Controller
Voltage 220 V
Width (English) 48 in.
Width (Metric) 121.92 cm