Learn More
Syngene™ Airclean 600 Dead Air Box
Product Code. 16584331 Shop All Syngene Products
Change view
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Unit Size
16584331 32 in. Each
16554341 48 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16584331 Supplier Syngene™ Supplier No. SYN32DBCE
  • ONLINE EXCLUSIVE PRICE
    76245.00 NOK
    67322.89 NOK / Pack of 1
    Save 8922.11 NOK
    11% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Airclean 600 dead air box equipped with a UVTect Controller.
  • With UVTect Controller
  • Price includes fluorescent lights, UV lights, UV shelf with pipette holder, and slip hatch
Specifications
Product Type Dead Air Box
Certifications/Compliance AC UL, CSA, and CE approved
Includes UV Controller
Voltage 220 V
Width (English) 32 in.
Width (Metric) 81.28 cm