Learn More
Beskrivning
- Asecos PURIFIAIR.488 can be used to reduce pollutant levels in indoor atmospheres
- Removes mould spores, pollen, and allergens
- Eliminates germs and bacteria and neutralises them
- Continuously filters out fine dust particles
- Removes unpleasant odours and harmful gases from indoor atmospheres
- Built-in PM2.5 sensor and colour display allow users to quickly identify and easily read off the particle concentration in the indoor atmosphere in four levels from green (low) to red (high)
- 1 device handles a volume flow rate of up to 488 m3/h and is designed for approx. 80 m3 room volume (corresponding to a surface area of 32 m2; at a room height of 2.5 m)
- A larger number of devices should be used in larger rooms
- Fresh air must be supplied to the rooms via natural or technical ventilation
- Air purifiers should not be considered a substitute for a fresh air supply
- Efficient results over time only if the filter is replaced at regularly according to the electronic display
- Large filter units ensure a long service life
- Low energy requirements result in low usage costs
- Integrated humidification for a healthy and comfortable indoor climate
Specifikationer
Specifikationer
| Filter Type | Prefilter, Cold Catalysis Filter, Activated Carbon Filter, High-performance Particle Filter |
| Airflow Rate | 488 3/h |
| Weight (Metric) | 11.3 Kg |
| Height (Metric) | 669 mm |
| Depth (Metric) | 400 mm |
| Width (Metric) | 400 mm |
| Product Type | Air Purifier |
| Voltage | 220/240 V |
| Frequency | 50/60 Hz |
| Certifications/Compliance | CE Conformity |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?