Learn More
Cole-Parmer™ Air Lubricator
Shop All Cole-Parmer Products
Click to view available options
Thread Size:
1/4 in. NPT(F)
Unit Size:
Each
Description
- Air lubricator
- Uses light spindle oil
- Porous bronze wick
- Measures 4 1/4 x 1 5/8 in. dia.
Specifications
Specifications
| Description | 1 to 20 scfm |
| Port Size | 1/4 in. NPT(F) |
| Thread Size | 1/4 in. NPT(F) |
| Product Type | Air lubricator |