Learn More
Aalborg™ Air Direct Glass Flow Tube 150mm Reference Scale
Product Code. p-8490007
Click to view available options
Airflow Rate:
374 mL/min.
Unit Size:
Each
Product Code. 11788258
Brand: Aalborg™ FT11202SAVA
Only null left
Add to basket
Add to basket
Aalborg™ Air Direct Glass Flow Tube Reference Scale reults in superior mL and L readings by providing a scale of meaurements in a vairety of volumes.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Type Air Direct Flow Tube
For Use With (Application) For various fluids, gases or liquids, thanks to the correlation tables
Width (Metric) 150 mm
Airflow Rate 374 mL/min.