Learn More
Abeba™ Air-Cushion 32600 Shoes
Product Code. 15619727
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. Size unitSize
15619727 42 1 pair
15649717 35 1 pair
15659717 36 1 pair
15669717 37 1 pair
15679717 38 1 pair
15689717 39 1 pair
15699717 40 1 pair
15609727 41 1 pair
15629727 43 1 pair
15639727 44 1 pair
15649727 45 1 pair
15659727 46 1 pair
15669727 47 1 pair
13 options
Product Code. 15619727
Brand: Abeba™ 32600/42
Only null left
Add to basket
Add to basket
Air cushion PU-sole with high slip resistance. Sanitized™ treated.
  • CE EN ISO 20347:2012 O2 SRA
  • Grained leather
  • White
  • Slip resistance PU outsole sole
  • Replaceable insole (article no. 3551)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O2 SRA
Color White
Liner Material Microfiber
Material Leather
Size 42
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32600