Learn More
AIM2 Mouse anti-Human, Unconjugated, Clone: 3E12A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66902-1-IG-150UL
Description
The AIM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AIM2. This antibody recognizes human, mouse, rat, rabbit antigen. The AIM2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| AIM2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit |
| Western Blot | |
| 3E12A8 | |
| Unconjugated | |
| 150 μL | |
| 9447 | |
| IgG1 |