missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIFM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14778-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. Recently a protein similar to the apoptosis-inducing factor (AIF) was cloned and designated AIFL (also known as AIFM3). AIFM3 is expressed ubiquitously and is predomitly localized to the inner membranes of mitochondria. Unlike AIF, AIFM3 does not translocate to the nucleus upon induction of apoptosis. However, overexpression of AIFM3, like AIF, induced cytochrome c release from the mitochondria, cleavage of caspase 3, and ultimately apoptosis, indicating AIFM3 induces apoptosis through caspase activation. Multiple isoforms of AIFM3 are known to exist.Specifications
AIFM3 | |
Polyclonal | |
Unconjugated | |
AIFM3 | |
AIFL, AIFM3, Apoptosis inducing factor 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
150209, 303786, 72168 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TY86, Q96NN9 | |
Aifm3 | |
AIFM3 Fusion Protein Ag6278 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |