missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AHSG (Human) Recombinant Protein
Human AHSG full-length ORF ( AAH48198.1, 19 a.a. - 367 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000197-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: APHGPGLIYRQPNCDDPETEEAALVAIDYINQNLPWGYKHTLNQIDEVKVWPQQPSGELFEIEIDTLETTCHVLDPTPVARCSVRQLKEHAVEGDCDFQLLKLDGKFSVVYAKCDSSPDSAEDVRKVCQDCPLLAPLNDTRVVHAAKAALAAFNAQNNGSNFQLEEISRAQLVPLPPSTYVEFTVSGTDCVAKEATEAAKCNLLAEKQYGFCKATLSEKLGGAEVAVTCTVFQTQPVTSQPQPEGANEAVPTPVVDPDAPPSPPLGAPGLPPAGSPPDSHVLLAAPPGHQLHRAHYDLRHTFMGVVSLGSPSGEVSHPRKTRTVVQPSVGAAAGPVVPPCPGRIRHFKV
Specifications
AAH48198.1 | |
alpha-2-HS-glycoprotein | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AHSG | |
GST |
197 | |
Wheat germ expression system | |
25 μg | |
A2HS, AHS, FETUA, HSGA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |