Learn More
AHNAK2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17682-1-AP-20UL
This item is not returnable.
View return policy
Description
The specific function of AHNAK2 is not yet known.Specifications
| AHNAK2 | |
| Polyclonal | |
| Unconjugated | |
| AHNAK2 | |
| AHNAK nucleoprotein 2, AHNAK2, C14orf78, KIAA2019, Protein AHNAK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 113146 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IVF2 | |
| AHNAK2 | |
| AHNAK2 Fusion Protein Ag11779 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |