Learn More
AGER Mouse anti-Human, Unconjugated, Clone: 3D4G11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66833-1-IG-20UL
 View more versions of this product
Product Code. 16849075
  • 1710.33 NOK / 20µL
Description
The AGER antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AGER. This antibody recognizes human, pig, rat antigen. The AGER antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
AGER
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Pig
Western Blot, Immunohistochemistry
3D4G11
Unconjugated
20 μL
177
IgG1