Learn More
AGER Mouse anti-Human, Unconjugated, Clone: 3D4G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66833-1-IG-20UL
This item is not returnable.
View return policy
Description
The AGER antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AGER. This antibody recognizes human, pig, rat antigen. The AGER antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| AGER | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Pig |
| Western Blot, Immunohistochemistry | |
| 3D4G11 | |
| Unconjugated | |
| 20 μL | |
| 177 | |
| IgG1 |