Learn More
Agencinox™ Agencinox Stainless Steel Micro Spatula
Product Code. 11844592
Change view
Click to view available options
Length (Metric):
130 mm
Unit Size:
Pack of 5
Product Code. Length (Metric) unitSize
11844592 130 mm Pack of 5
1 options
Product Code. 11844592
Brand: Agencinox™ 1826
Only null left
Add to basket
Add to basket
Transfer your samples without contamination, sticking and the assurance of complete release. Agencinox™ Stainless Steel Micro Spatulas are chemically inert and manufactured with smooth, flat surfaces.
  • Stainless steel micro spatula
  • Ideal for most laboratory applications
Specifications
Quantity 5 Pk.
Certifications/Compliance CE, ISO
Product Type Micro Spatula
Material Stainless Steel
Length (Metric) 130 mm