Learn More
RETSCH Agate Hand Mortar
Shop All Retsch Products
Click to view available options
Capacity (Metric):
10 mL
30 mL
130 mL
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Diameter (Metric) Inner | 65mm |
| Diameter (Metric) Outer | 80mm |
| Product Type | Hand Mortar |
| Material | Agate Stone |
| For Use With (Application) | Suitable for grinding, mixing and triturating medium-hard to hard substances |
| Type | Hand Mortar |