Learn More
RETSCH Agate Grinding Jars
Product Code. 10228230 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
For MM 200 Mixer Mill
Capacity (Metric):
5 mL
10 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Capacity (Metric) Unit Size
10228230 For MM 200 Mixer Mill 5 mL Each
10771581 For MM 200 Mixer Mill 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10228230 Supplier RETSCH Supplier No. 01.462.0113
  • ONLINE EXCLUSIVE PRICE
    10630.00 NOK
    9675.61 NOK / Pack of 1
    Save 954.39 NOK
    8% Off
Estimated Shipment: 07-12-2026
Use these grinding jars for dry, wet and cryogenic grinding of small samples. Retsch™ Agate Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
  • Screw-top grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Product Type Grinding Jar
Capacity (Metric) 5 mL
For Use With (Equipment) For MM 200 Mixer Mill
Material Agate