Learn More
RETSCH Agate Grinding Balls
Brand: RETSCH 05.368.0065
4683.50 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.03500kg
Specifications
Grinding Balls | |
30 mm | |
Planetary Ball Mill PM 400 | |
Agate |