Learn More
SPEX™ Agate Grinding Ball
SPEX™ Agate Grinding Ball is a replacement ball for use with 8014 and 3118 Agate Grinding Vial Set.
Brand:  SPEX™ 8014A
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 12558706
  • 585.00 NOK / Each
Estimated Shipment: 29-05-2024
Description
  • Ideal for grinding and mixing samples when organic and metallic contamination are equally undesirable
  • Extremely high resistance to abrasion and very long wearing withmoderate efficiency
  • Handmade from flawless Brazilian agate
  • Typical composition: Major element – Si; Minor elements – Al, Na, Fe, K, Ca, Mg
  • Hardness: Mohs 6-7; Knoop 550-800
Specifications
Specifications
Grinding Balls
12.7 mm
8014 Agate Grinding Vial Set
Agate