Learn More
Thermo Scientific Chemicals Agarose medium-low EEO, for electrophoresis, (Mr = 0.12)
Product Code. p-30087 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
Product Code. 10289590
Brand: Thermo Scientific Chemicals undefined
Request Bulk or Custom Format
CAS: 9012-36-6
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9012-36-6
MDL Number MFCD00081294
Specifications
Physical Form Fine Crystalline Powder
Melting Point 86.5°C to 89.5°C (1.5% gel)
Color Cream to White
Quantity 25 g
Electroendosmosis (EEO) 0.05 to 0.13
Gelation Temperature 34.5° to 37.5°C (1.5% gel)
Gel Strength >1200g/cm2 (1% gel), >2500g/cm2 (1.5% gel)
Infrared Spectrum Authentic
Packaging Glass Bottle
Grade Electrophoresis
Chemical Name or Material Agarose medium-low EEO