Learn More
Agarose gel loading dye (6X), sucrose based, Thermo Scientific™
Product Code. 15413119
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
Product Code. 15413119
Brand: Thermo Scientific Alfa Aesar J60333.AD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Agarose gel loading dye
Name Note sucrose based
Physical Form Liquid
Concentration 6X
Quantity 50 mL