Learn More
Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Ficoll-Based)
Product Code. 15485099
Change view
Click to view available options
Quantity:
10 mL
25 mL
Unit Size:
10mL
25mL
Product Code. Quantity unitSize
15485099 25 mL 25mL
15475099 10 mL 10mL
2 options
Product Code. 15485099
Brand: Thermo Scientific Chemicals J62800.AC
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Agarose gel loading dye
Name Note Ficoll based
Physical Form Viscous Liquid
Concentration 6X
Quantity 25 mL
MDL Number MFCD00675058