missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Agar Technical No.2

Product Code. p-200112595
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15562141

Brand: Thermo Scientific™ LP0012B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Agar Technical (Agar No. 2) is a general-purpose bacteriological-grade agar.

A technical grade agar which conforms to the specifications of the U.S. Pharmacopoeia and A.P.H.A. Suitable for applications where clarity and compatibility are not of prime importance. It is of high gel strength and will give a firm gel 1.2% w/v.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Agar
Fill Weight 500 g
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.