Learn More
Thermo Scientific Chemicals Agar, pure, powder
Product Code. p-4258662 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
25 kg
Unit Size:
1kg
250g
25kg
500g
5kg
Product Code. Quantity unitSize
10782453 250 g 250g
10048991 1 kg 1kg
10465872 5 kg 5kg
11946991 25 kg 25kg
10589462 500 g 500g
5 options
Product Code. 10782453
Brand: Thermo Scientific Chemicals 400402500
Request Bulk or Custom Format
For use in biological culture media.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Product Type Agar
Packaging Type Glass Bottle
Quantity 250 g