missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ African Swine Fever Virus p30 (Ba71V-93) Monoclonal Antibody (2E1F3)
GREENER_CHOICE

Product Code. 17940053
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17940053 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17940053 Supplier Invitrogen™ Supplier No. MA550275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

African Swine Fever Virus p30 (Ba71V-93) Monoclonal Antibody for ELISA

African swine fever virus (ASFV) is a complex nucleocytoplasmic large DNA virus (NCLDV) that causes a lethal hemorrhagic disease that is currently threatening the global pig industry. ASFV structural protein p30 is a membrane phosphoprotein that suggests it may play a regulatory role, possibly in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen African Swine Fever Virus p30 (Ba71V-93)
Applications ELISA, Neutralization
Classification Monoclonal
Clone 2E1F3
Concentration 2.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Ba71V-93
Gene Accession No. P34204
Gene Alias Ba71V-93; CP204L; Phosphoprotein p30
Gene Symbols Ba71V-93
Host Species Mouse
Immunogen Recombinant Phosphoprotein p30 protein (1-204AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22220322
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.