missing translation for 'onlineSavingsMsg'
Learn More

AFN-1252, MedChemExpress

Product Code. p-200095930
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16463227 50 mg 50mg
16457208 10 mg 10mg
16449737 5 mg 5mg
3 options
This item is not returnable. View return policy

Product Code. 16463227

Brand: Medchem Express HY16911

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AFN-1252(Debio 1452) is a potent inhibitor of enoyl-acyl carrier protein reductase (FabI), inhibited all clinical isolates of Staphylococcus aureus and Staphylococcus epidermidis at concentrations of ≤0.12 μg/ml.

AFN-1252(Debio 1452) is a potent inhibitor of enoyl-acyl carrier protein reductase (FabI), inhibited all clinical isolates of Staphylococcus aureus and Staphylococcus epidermidis at concentrations of ≤0.12 μg/ml.

Specifications

Chemical Name or Material AFN-1252
Physical Form Solid
CAS 620175-39-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H21N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym API-1252 Debio 1452
Solubility Information DMSO : 5.8 mg/mL (15.45 mM; Need ultrasonic)
SMILES O=C(N(C)CC1=C(C)C2=CC=CC=C2O1)/C=C/C(C=C3CC4)=CNC3=NC4=O
Molecular Weight (g/mol) 375.42
Formula Weight 375.42
Percent Purity 99.13%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.