Learn More
Applied Biosystems™ ExoSAP-IT™ For PCR Product Clean-Up
Product Code. 15593677
Change view
Click to view available options
Quantity:
10 mL
4 x 1 mL
Unit Size:
2000 reactions
5000 reactions
Product Code. Quantity unitSize
15593677 4 x 1 mL 2000 reactions
15503687 10 mL 5000 reactions
2 options
Product Code. 15593677
Brand: Applied Biosystems™ 78202.4X.1.ML
Edge
Designed for simple, quick PCR cleanup for downstream applications, such as DNA sequencing or Single Nucleotide Polymorphism (SNP) analysis. Single-step enzymatic cleanup of PCR products that eliminates unincorporated primers and dNTPs.
  • Two hydrolytic enzymes, Exonuclease I and Shrimp Alkaline Phosphatase (SAP), together in a specially formulated buffer
  • 30-minute protocol
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
For Use With (Application) Quick PCR clean-up for downstream applications; Eliminates unincorporated primers and dNTPs
No. of Reactions 2000
Product Type Single-step enzymatic PCR Cleanup Kit
Quantity 4 x 1 mL