Description
AffiniPure Polyclonal antibody specifically detects AffiniPure in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.
Specifications
Specifications
| Antigen | AffiniPure |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.32 mg/mL |
| Conjugate | HRP |
| Host Species | Goat |
| Quantity | 200 μL |
| Primary or Secondary | Secondary |
| Target Species | Human, Mouse, Rat |
| Form | Liquid |
| Show More |